2,2,3,3,3-Pentafluoropropyl trifluoromethanesulphonate 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1327-25G
PC1327-5G
PC1327-1KG
APOSPC1327-25GEA
9920
CZK
APOSPC1327-25G
APOSPC1327-5G
APOSPC1327-1KG
2,2,3,3,3-Pentafluoropropyl trifluoromethanesulphonate 95%
2,2,3,3,3-Pentafluoropropyl trifluoromethanesulphonate
Vzorec:
C₄H₂F₈O₃S MW: 282,11 g/mol Bod varu: 102…105 °C/740mmHg Hustota: 1,702 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01862006 CAS číslo: 6401-00-9 UN: 2927 ADR: 6.1+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...