3-(2-Fluorophenoxy)propionic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(2-Fluorophenoxy)propionic Acid
PC1317-1G
PC1317-250MG
PC1317-5G
APOSPC1317-1GEA
5820
CZK
APOSPC1317-1G
APOSPC1317-250MG
APOSPC1317-5G
3-(2-Fluorophenoxy)propionic acid
3-(2-Fluorophenoxy)propionic acid
Vzorec:
C₉H₉FO₃ MW: 184.1600 g/mol Bod tání: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569309 CAS číslo: 2967-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...