2,2,2-Trifluoroethyl nonafluorobutanesulphonate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,2,2-Trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulphonate, 2,2,2-Trifluoroethyl Perfluorobutanesulphonate, Nonafluorobutanesulphonic acid 2,2,2-Trifluoroethyl Ester, Perfluorobutanesulphonic acid 2,2,2-Trifluoroethyl Ester
PC1302-100G
PC1302-25G
APOSPC1302-100GEA
14290
CZK
APOSPC1302-100G
APOSPC1302-25G
2,2,2-Trifluoroethyl nonafluorobutanesulphonate 97%
2,2,2-Trifluoroethyl nonafluorobutanesulphonate
Vzorec:
C₆H₂F₁₂O₃S MW: 382,12 g/mol Bod varu: 143…144 °C Hustota: 1,636 g/cm³ (25 °C) Bod vzplanutí: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093638 CAS číslo: 79963-95-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...