Potassium-2,2,3,3-tetrafluoropropionate 97%
Dodavatel: Apollo Scientific
PC1299-100MG
PC1299-1G
PC1299-250MG
APOSPC1299-100MGEA
4770
CZK
APOSPC1299-100MG
APOSPC1299-1G
APOSPC1299-250MG
Potassium-2,2,3,3-tetrafluoropropionate 97%
Potassium-2,2,3,3-tetrafluoropropionate
Vzorec:
C₃HF₄KO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03094125 CAS číslo: 71592-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...