Tisk…

2-(2-Hydroxy-1,1,1,3,3,3-hexafluoropropyl)-1-naphthol 97%

Dodavatel: Apollo Scientific

PC1282-1G PC1282-25G PC1282-5G
APOSPC1282-1GEA 3940 CZK
APOSPC1282-1G APOSPC1282-25G APOSPC1282-5G
2-(2-Hydroxy-1,1,1,3,3,3-hexafluoropropyl)-1-naphthol 97%
2-(2-Hydroxy-1,1,1,3,3,3-hexafluoropropyl)-1-naphthol
Vzorec: C₁₃H₈F₆O₂
MW: 310,2 g/mol
Bod tání: 119…120 °C
Storage Temperature: Ambient
MDL Number: MFCD06247736
CAS číslo: 2092-87-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR