Tisk…

3-Chloro-2-iodo-1,1,1-trifluoropropane 97%

Dodavatel: Apollo Scientific
Varování

PC1268-25G PC1268-5G
APOSPC1268-25GEA 12240 CZK
APOSPC1268-25G APOSPC1268-5G
3-Chloro-2-iodo-1,1,1-trifluoropropane 97%
3-Chloro-2-iodo-1,1,1-trifluoropropane
Vzorec: C₃H₃ClF₃I
Storage Temperature: Ambient
MDL Number: MFCD01862016
CAS číslo: 113402-77-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR