Tisk…

Bis(4-chlorooctafluorobutyl)ether 97%

Dodavatel: Apollo Scientific

PC1261-25G PC1261-5G
APOSPC1261-25GEA 19080 CZK
APOSPC1261-25G APOSPC1261-5G
Bis(4-chlorooctafluorobutyl)ether 97%
Bis(4-chlorooctafluorobutyl)ether
Vzorec: C₈Cl₂F₁₆O
MW: 486.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094104
CAS číslo: 149697-40-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR