Bis(2,2,2-trifluoroethyl)amine 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1253-1G
PC1253-5G
PC1253-25G
APOSPC1253-1GEA
1820
CZK
APOSPC1253-1G
APOSPC1253-5G
APOSPC1253-25G
Bis(2,2,2-trifluoroethyl)amine 95%
Bis(2,2,2-trifluoroethyl)amine
Vzorec:
C₄H₅F₆N Bod varu: 70…71 °C Hustota: >1,1 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042094 CAS číslo: 407-01-2 EINECS: 206-978-7 UN: 2924 ADR: 3+8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...