Tisk…

(Pentafluoroethyl)trimethylsilane 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC1217-1G PC1217-25G PC1217-5G
APOSPC1217-1GEA 1590 CZK
APOSPC1217-1G APOSPC1217-25G APOSPC1217-5G
(Pentafluoroethyl)trimethylsilane 97%
(Pentafluoroethyl)trimethylsilane
Vzorec: C₅H₉F₅Si
Bod varu: 60…70 °C
Hustota: 1,11 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04116436
CAS číslo: 124898-13-1
UN: 1993
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR