4-Bromo-2,6-difluorophenol 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Bromo-3,5-difluoro-4-hydroxybenzene
PC1209-1G
PC1209-5G
PC1209-10G
APOSPC1209-1GEA
622
CZK
APOSPC1209-1G
APOSPC1209-5G
APOSPC1209-10G
4-Bromo-2,6-difluorophenol 98%
4-Bromo-2,6-difluorophenol
Vzorec:
C₆H₃BrF₂O MW: 208,99 g/mol Bod tání: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236213 CAS číslo: 104197-13-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...