Tisk…

3,4-Difluoro-5-methoxyphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (3,4-Difluoro-5-methoxyphenyl)boronic acid

PC1202-1G PC1202-25G PC1202-5G
APOSPC1202-1GEA 3380 CZK
APOSPC1202-1G APOSPC1202-25G APOSPC1202-5G
3,4-Difluoro-5-methoxyphenylboronic acid
3,4-Difluoro-5-methoxyphenylboronic acid
Vzorec: C₇H₇BF₂O₃
MW: 187,940 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09907960
CAS číslo: 925910-42-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR