2-Fluoroisonicotinaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Fluoropyridine-4-carboxaldehyde
PC1186-1G
PC1186-250MG
APOSPC1186-1GEA
4570
CZK
APOSPC1186-1G
APOSPC1186-250MG
2-Fluoroisonicotinaldehyde
2-Fluoroisonicotinaldehyde
Vzorec:
C₆H₄FNO MW: 125,1000 g/mol Bod varu: 52 °C/0,53mmHg Storage Temperature: Freezer |
MDL Number:
MFCD06410677 CAS číslo: 131747-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...