4-Amino-2-chloro-5-fluoropyrimidine
Dodavatel: Apollo Scientific
|
Varování
|
PC1168-1G
PC1168-5G
APOSPC1168-1GEA
1740
CZK
APOSPC1168-1G
APOSPC1168-5G
4-Amino-2-chloro-5-fluoropyrimidine
4-Amino-2-chloro-5-fluoropyrimidine
Vzorec:
C₄H₃ClFN₃ MW: 147,54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057344 CAS číslo: 155-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...