2-Bromo-2'-chloro-4'-fluoroacetophenone
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1151-1G
PC1151-250MG
PC1151-5G
APOSPC1151-1GEA
1910
CZK
APOSPC1151-1G
APOSPC1151-250MG
APOSPC1151-5G
2-Bromo-2'-chloro-4'-fluoroacetophenone
2-Bromo-2'-chloro-4'-fluoroacetophenone
Vzorec:
C₈H₅BrClFO Storage Temperature: Refrigerator |
MDL Number:
MFCD08059508 CAS číslo: 61397-54-4 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...