3,6-Difluorosalicylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,6-Difluoro-2-hydroxybenzoic acid
PC1139-1G
PC1139-500MG
PC1139-5G
APOSPC1139-1GEA
3110
CZK
APOSPC1139-1G
APOSPC1139-500MG
APOSPC1139-5G
3,6-Difluorosalicylic acid
3,6-Difluorosalicylic acid
Vzorec:
C₇H₄F₂O₃ MW: 174,10 g/mol Bod tání: 154…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275285 CAS číslo: 749230-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...