Tisk…

Pentafluorophenyl-3,4-dimethoxybenzenesulphonate

Dodavatel: Apollo Scientific

PC11273-1G PC11273-500MG PC11273-5G
APOSPC11273-1GEA 7840 CZK
APOSPC11273-1G APOSPC11273-500MG APOSPC11273-5G
Pentafluorophenyl-3,4-dimethoxybenzenesulphonate
Pentafluorophenyl-3,4-dimethoxybenzenesulphonate
Vzorec: C₁₄H₉F₅O₅S
Bod tání: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD05975142
CAS číslo: 886361-12-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR