2-Chloro-N-(2,5-difluorophenyl)acetamide
Dodavatel: Apollo Scientific
Synonyms:
N-Chloroacetyl-2,5-difluoroaniline
PC11251-10G
PC11251-1G
PC11251-5G
APOSPC11251-10GEA
7440
CZK
APOSPC11251-10G
APOSPC11251-1G
APOSPC11251-5G
2-Chloro-N-(2,5-difluorophenyl)acetamide
2-Chloro-N-(2,5-difluorophenyl)acetamide
Vzorec:
C₈H₆ClF₂NO MW: 205,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00157387 CAS číslo: 196938-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...