Potassium-4-carboxyphenyltrifluoroborate
Dodavatel: Apollo Scientific
Synonyms:
Potassium 4-carboxyphenyltrifluoroborate, Potassium (4-carboxyphenyl)trifluoroborate
PC11245-100G
PC11245-25G
PC11245-5G
APOSPC11245-100GEA
33730
CZK
APOSPC11245-100G
APOSPC11245-25G
APOSPC11245-5G
Potassium-4-carboxyphenyltrifluoroborate
Potassium-4-carboxyphenyltrifluoroborate
Vzorec:
C₇H₅BF₃KO₂ MW: 228,02 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115737 CAS číslo: 850623-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...