Potassium(3-nitrophenyl)trifluoroborate
Dodavatel: Apollo Scientific
Synonyms:
(3-Nitrophenyl)trifluoroboric acid potassium salt, Potassium trifluoro(3-nitrophenyl)borate
PC11242-100G
PC11242-25G
PC11242-5G
APOSPC11242-100GEA
29830
CZK
APOSPC11242-100G
APOSPC11242-25G
APOSPC11242-5G
Potassium(3-nitrophenyl)trifluoroborate
Potassium(3-nitrophenyl)trifluoroborate
Vzorec:
C₆H₄BF₃KNO₂ MW: 229,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03701627 CAS číslo: 192863-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...