Tisk…

Potassium (3-methylphenyl)trifluoroborate

Dodavatel: Apollo Scientific

Synonyms: Potassium trifluoro(m-tolyl)borate, (3-Methylphenyl)trifluoroboric acid potassium salt

PC11241-25G PC11241-5G
APOSPC11241-25GEA 14920 CZK
APOSPC11241-25G APOSPC11241-5G
Potassium (3-methylphenyl)trifluoroborate
Potassium (3-methylphenyl)trifluoroborate
Vzorec: C₇H₇BF₃K
MW: 198,04 g/mol
Bod tání: 226…230 °C
Storage Temperature: Ambient
MDL Number: MFCD04115739
CAS číslo: 850623-42-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR