Potassium-4-formylphenyltrifluoroborate 98%
Dodavatel: Apollo Scientific
Synonyms:
Potassium (4-formylphenyl)trifluoroborate, Potassium trifluoro(4-formylphenyl)borate
PC11237-100G
PC11237-25G
APOSPC11237-100GEA
15770
CZK
APOSPC11237-100G
APOSPC11237-25G
Potassium-4-formylphenyltrifluoroborate 98%
Potassium-4-formylphenyltrifluoroborate
Vzorec:
C₇H₅BF₃KO Bod tání: 282…288 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093980 CAS číslo: 374564-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...