Potassium-3-(methylthio)phenyltrifluoroborate
Dodavatel: Apollo Scientific
PC11236-100G
PC11236-25G
PC11236-5G
APOSPC11236-100GEA
22390
CZK
APOSPC11236-100G
APOSPC11236-25G
APOSPC11236-5G
Potassium-3-(methylthio)phenyltrifluoroborate
Potassium-3-(methylthio)phenyltrifluoroborate
Vzorec:
C₇H₇BF₃KS Bod tání: 70…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115747 CAS číslo: 850623-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...