Tisk…

Potassium(4-methyl-1-naphthalene)trifluoroborate

Dodavatel: Apollo Scientific

PC11235-100G PC11235-25G PC11235-5G
APOSPC11235-100GEA 22390 CZK
APOSPC11235-100G APOSPC11235-25G APOSPC11235-5G
Potassium(4-methyl-1-naphthalene)trifluoroborate
Potassium (4-methyl-1-naphthalene)trifluoroborate
Vzorec: C₁₁H₉BF₃K
MW: 248,09 g/mol
Bod tání: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD04115748
CAS číslo: 850623-55-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR