Potassium(4-ethoxyphenyl)trifluoroborate
Dodavatel: Apollo Scientific
Synonyms:
Potassium (4-ethoxyphenyl)trifluoroborate, (4-Ethoxyphenyl)trifluoroboric acid potassium salt
PC11230-1G
PC11230-25G
PC11230-5G
APOSPC11230-1GEA
3440
CZK
APOSPC11230-1G
APOSPC11230-25G
APOSPC11230-5G
Potassium(4-ethoxyphenyl)trifluoroborate
Potassium(4-ethoxyphenyl)trifluoroborate
Vzorec:
C₈H₉BF₃KO MW: 228,06 g/mol Bod tání: 298…304 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115753 CAS číslo: 850623-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...