Tisk…

Potassium (3-(methanesulphonylamino)phenyl)trifluoroborate

Dodavatel: Apollo Scientific

Synonyms: Potassium trifluoro(3-(methylsulfonamido)phenyl)borate

PC11228-10G PC11228-1G PC11228-5G
APOSPC11228-10GEA 37360 CZK
APOSPC11228-10G APOSPC11228-1G APOSPC11228-5G
Potassium (3-(methanesulphonylamino)phenyl)trifluoroborate
Potassium (3-(methanesulphonylamino)phenyl)trifluoroborate
Vzorec: C₇H₈BF₃KNO₂S
MW: 277,11 g/mol
Bod tání: 180…190 °C
Storage Temperature: Ambient
MDL Number: MFCD04115762
CAS číslo: 850623-67-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR