Tisk…

Potassium-2-(methylsulphonyl)phenyltrifluoroborate

Dodavatel: Apollo Scientific

Synonyms: Potassium trifluoro(2-(methylsulphonyl)phenyl)borate, Potassium (2-methylsulphonylphenyl)trifluoroborate

PC11227-1G PC11227-25G PC11227-5G
APOSPC11227-1GEA 3940 CZK
APOSPC11227-1G APOSPC11227-25G APOSPC11227-5G
Potassium-2-(methylsulphonyl)phenyltrifluoroborate
Potassium-2-(methylsulphonyl)phenyltrifluoroborate
Vzorec: C₇H₇BF₃KO₂S
Bod tání: 78…84 °C
Storage Temperature: Ambient
MDL Number: MFCD04115760
CAS číslo: 850623-65-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR