Potassium (2-nitrophenyl)trifluoroborate 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Potassium trifluoro(2-nitrophenyl)borate, (2-Nitrophenyl)trifluoroboric acid potassium salt
PC11226-1G
PC11226-5G
PC11226-25G
APOSPC11226-1GEA
4010
CZK
APOSPC11226-1G
APOSPC11226-5G
APOSPC11226-25G
Potassium (2-nitrophenyl)trifluoroborate 96%
Potassium (2-nitrophenyl)trifluoroborate
Vzorec:
C₆H₄BF₃KNO₂ MW: 229.01 Bod tání: 216…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115759 CAS číslo: 850623-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...