Tisk…

Potassium(4-methoxyphenyl)trifluoroborate

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Methoxyphenyl)trifluoroboric acid potassium salt, Potassium (4-methoxyphenyl)trifluoroborate, Potassium trifluoro(4-methoxyphenyl)borate

PC11225-25G PC11225-5G
APOSPC11225-25GEA 7910 CZK
APOSPC11225-25G APOSPC11225-5G
Potassium(4-methoxyphenyl)trifluoroborate
Potassium(4-methoxyphenyl)trifluoroborate
Vzorec: C₇H₇BF₃KO
Storage Temperature: Ambient
MDL Number: MFCD04112717
CAS číslo: 192863-36-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR