2,4-Dichloro-5-fluoro-3-nitrobenzoic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC11216-100G
PC11216-25G
APOSPC11216-100GEA
2400
CZK
APOSPC11216-100G
APOSPC11216-25G
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid 99%
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid
Vzorec:
C₇H₂Cl₂FNO₄ MW: 254 g/mol Bod tání: 190…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427137 CAS číslo: 106809-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...