4'-(Trifluoromethyl)biphenyl-4-carboxylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid
PC11180-1G
PC11180-5G
APOSPC11180-1GEA
2150
CZK
APOSPC11180-1G
APOSPC11180-5G
4'-(Trifluoromethyl)biphenyl-4-carboxylic acid 98%
4'-(Trifluoromethyl)biphenyl-4-carboxylic acid
Vzorec:
C₁₄H₉F₃O₂ MW: 266.22 Bod tání: 321…324 °C Storage Temperature: Ambient |
MDL Number:
MFCD03424606 CAS číslo: 195457-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...