3'-(Trifluoromethyl)biphenyl-4-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid
PC11178-1G
PC11178-250MG
PC11178-5G
APOSPC11178-1GEA
3340
CZK
APOSPC11178-1G
APOSPC11178-250MG
APOSPC11178-5G
3'-(Trifluoromethyl)biphenyl-4-carboxylic acid
3'-(Trifluoromethyl)biphenyl-4-carboxylic acid
Vzorec:
C₁₄H₉F₃O₂ MW: 266,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03424607 CAS číslo: 195457-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...