Tisk…

Diflunisal

Dodavatel: Apollo Scientific
Varování

Synonyms: 2',4'-Difluoro-4-hydroxy[1,1'-biphenyl]-3-carboxylic acid, 2',4'-Difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid

PC11123-100G PC11123-25G PC11123-5G
APOSPC11123-100GEA 7180 CZK
APOSPC11123-100G APOSPC11123-25G APOSPC11123-5G
Diflunisal
Diflunisal
Vzorec: C₁₃H₈F₂O₃
MW: 250,1976 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057834
CAS číslo: 22494-42-4
EINECS: 245-034-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR