Tisk…

4-(Trifluoromethyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

PC1111-100G PC1111-25G PC1111-5G
APOSPC1111-100GEA 3110 CZK
APOSPC1111-100G APOSPC1111-25G APOSPC1111-5G
4-(Trifluoromethyl)phenylboronic acid 98%
4-(Trifluoromethyl)phenylboronic acid
Vzorec: C₇H₆BF₃O₂
MW: 189,93 g/mol
Bod tání: 235…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00151855
CAS číslo: 128796-39-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR