Tisk…

Ammonium tetrafluoroborate 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

PC1110-100G PC1110-25G PC1110-500G
APOSPC1110-100GEA 705 CZK
APOSPC1110-100G APOSPC1110-25G APOSPC1110-500G
Ammonium tetrafluoroborate 98%
Ammonium tetrafluoroborate
Vzorec: H₄BF₄N
Bod tání: 230 °C
Hustota: 1,871 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06245785
CAS číslo: 13826-83-0
EINECS: 237-531-4
UN: 3260
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR