Triammonium hexafluoroaluminate 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Ammonium hexafluoroaluminate
PC1101-100G
PC1101-500G
APOSPC1101-100GEA
7480
CZK
APOSPC1101-100G
APOSPC1101-500G
Triammonium hexafluoroaluminate 99%
tri-Ammonium hexafluoroaluminate
Vzorec:
H₁₂AlF₆N₃ MW: 195,09 g/mol Bod varu: 239,5 °C Bod tání: 126,1 °C Hustota: 1,78 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00042144 CAS číslo: 7784-19-2 EINECS: 232-052-7 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...