3-Amino-4-(4-morpholino)benzotrifluoride 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(4-Morpholinyl)-5-(trifluoromethyl)aniline, 2-Morpholino-5-(trifluoromethyl)aniline
PC1100D-1G
PC1100D-250MG
APOSPC1100D-1GEA
787
CZK
APOSPC1100D-1G
APOSPC1100D-250MG
3-Amino-4-(4-morpholino)benzotrifluoride 97%
3-Amino-4-(4-morpholino)benzotrifluoride
Vzorec:
C₁₁H₁₃F₃N₂O MW: 246,23 g/mol Bod tání: 127…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042160 CAS číslo: 784-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...