Tisk…

(±)-3-Amino-4,4,4-trifluorobutyric acid 97%

Dodavatel: Apollo Scientific
Varování

PC1100B-1G PC1100B-5G
APOSPC1100B-1GEA 1740 CZK
APOSPC1100B-1G APOSPC1100B-5G
(±)-3-Amino-4,4,4-trifluorobutyric acid 97%
(±)-3-Amino-4,4,4-trifluorobutyric acid
Vzorec: C₄H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00041413
CAS číslo: 584-20-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR