Tisk…

2-Fluoro-5-(trifluoromethoxy)benzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC1095-1G PC1095-5G
APOSPC1095-1GEA 6620 CZK
APOSPC1095-1G APOSPC1095-5G
2-Fluoro-5-(trifluoromethoxy)benzyl bromide
2-Fluoro-5-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₅BrF₄O
MW: 273.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115890
CAS číslo: 86256-24-8
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR