Tisk…

2-Nitro-4-(trifluoromethyl)aniline 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Nitro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-nitrobenzotrifluoride, 4-Amino-3-nitro-α,α,α-trifluorotoluene

PC1090-1G PC1090-10G
APOSPC1090-1GEA 417 CZK
APOSPC1090-1G APOSPC1090-10G
2-Nitro-4-(trifluoromethyl)aniline 97%
2-Nitro-4-(trifluoromethyl)aniline
Vzorec: C₇H₅F₃N₂O₂
MW: 206,12 g/mol
Bod tání: 105…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007155
CAS číslo: 400-98-6
EINECS: 206-926-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR