4-Amino-3-fluorophenol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Fluoro-4-hydroxyaniline
PC1076J-25G
PC1076J-5G
APOSPC1076J-25GEA
992
CZK
APOSPC1076J-25G
APOSPC1076J-5G
4-Amino-3-fluorophenol
4-Amino-3-fluorophenol
Vzorec:
C₆H₆FNO MW: 127,12 g/mol Bod tání: 137…139 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077452 CAS číslo: 399-95-1 EINECS: 402-230-0 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...