Tisk…

2-Amino-6-fluorobenzylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Aminomethyl)-3-fluoroaniline

PC1076C-1G PC1076C-25G PC1076C-5G
APOSPC1076C-1GEA 2070 CZK
APOSPC1076C-1G APOSPC1076C-25G APOSPC1076C-5G
2-Amino-6-fluorobenzylamine 97%
2-Amino-6-fluorobenzylamine
Vzorec: C₇H₉FN₂
MW: 140.16 g/mol
Bod varu: 78…80 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD00042483
CAS číslo: 175277-93-7
UN: 2735
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR