5-Amino-2-fluorobenzoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC1071P-25G
PC1071P-5G
PC1071P-100G
APOSPC1071P-25GEA
1660
CZK
APOSPC1071P-25G
APOSPC1071P-5G
APOSPC1071P-100G
5-Amino-2-fluorobenzoic acid 97%
5-Amino-2-fluorobenzoic acid
Vzorec:
C₇H₆FNO₂ MW: 155,13 g/mol Bod tání: 190…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077449 CAS číslo: 56741-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...