Tisk…
Difluoromethyl phenyl sulphide 98%
Dodavatel: Apollo Scientific
PC10684-1G PC10684-5G
APOSPC10684-1GEA 2610 CZK
APOSPC10684-1G APOSPC10684-5G
Difluoromethyl phenyl sulphide 98%
Difluoromethyl phenyl sulphide
Vzorec: C₇H₆F₂S
MW: 160,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16301013
CAS číslo: 1535-67-7