2,3,4,5-Tetrafluorophenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3,4,5-Tetrafluorobenzeneboronic acid, Dihydroxy(2,3,4,5-tetrafluorophenyl)borane, (2,3,4,5-Tetrafluorophenyl)metaboric acid, (2,3,4,5-Tetrafluorophenyl)boronic acid
PC10540-1G
PC10540-250MG
APOSPC10540-1GEA
11410
CZK
APOSPC10540-1G
APOSPC10540-250MG
2,3,4,5-Tetrafluorophenylboronic acid 97%
2,3,4,5-Tetrafluorophenylboronic acid
Vzorec:
C₆H₃BF₄O₂ MW: 193,89 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04039308 CAS číslo: 179923-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...