3-Bromo-2,4,5-trifluorobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC10514-1G
PC10514-25G
PC10514-5G
APOSPC10514-1GEA
622
CZK
APOSPC10514-1G
APOSPC10514-25G
APOSPC10514-5G
3-Bromo-2,4,5-trifluorobenzoic acid
3-Bromo-2,4,5-trifluorobenzoic acid
Vzorec:
C₇H₂BrF₃O₂ MW: 254,99 g/mol Bod tání: 122…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664065 CAS číslo: 104222-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...