2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride
Dodavatel: Apollo Scientific
Synonyms:
2-Bromo-3,4,5,6-tetrafluorobenzene-1-carbonyl chloride
PC10511-5G
PC10511-25G
APOSPC10511-5GEA
4960
CZK
APOSPC10511-5G
APOSPC10511-25G
2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride
2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride
Vzorec:
C₇BrClF₄O MW: 291,43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001174 CAS číslo: 151096-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...