Tisk…

2-Bromo-4-fluorobenzoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-4-fluorobenzoylchloride

PC10444-1G PC10444-5G
APOSPC10444-1GEA 622 CZK
APOSPC10444-1G APOSPC10444-5G
2-Bromo-4-fluorobenzoyl chloride
2-Bromo-4-fluorobenzoyl chloride
Vzorec: C₇H₃BrClFO
MW: 237,46 g/mol
Bod varu: 108…109 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD07368700
CAS číslo: 95383-36-1
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR