3-Bromo-5-fluorobenzoyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC10443-1G
PC10443-5G
PC10443-25G
APOSPC10443-1GEA
622
CZK
APOSPC10443-1G
APOSPC10443-5G
APOSPC10443-25G
3-Bromo-5-fluorobenzoyl chloride
3-Bromo-5-fluorobenzoyl chloride
Vzorec:
C₇H₃BrClFO Bod varu: 88…90 °C/14mmHg Storage Temperature: Ambient |
MDL Number:
MFCD07368699 CAS číslo: 887266-90-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...