2-Bromo-3'-fluoro-4'-methylacetophenone
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Fluoro-4-methylphenacyl bromide
PC10429-1G
PC10429-250MG
PC10429-5G
APOSPC10429-1GEA
2980
CZK
APOSPC10429-1G
APOSPC10429-250MG
APOSPC10429-5G
2-Bromo-3'-fluoro-4'-methylacetophenone
2-Bromo-3'-fluoro-4'-methylacetophenone
Vzorec:
C₉H₈BrFO Bod tání: 62…64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07368693 CAS číslo: 505097-09-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...