Tisk…

2-Bromo-2',3',6'-trifluoroacetophenone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,3,6-Trifluorophenacyl bromide

PC10426-1G PC10426-5G PC10426-10G
APOSPC10426-1GEA 6950 CZK
APOSPC10426-1G APOSPC10426-5G APOSPC10426-10G
2-Bromo-2',3',6'-trifluoroacetophenone
2-Bromo-2',3',6'-trifluoroacetophenone
Vzorec: C₈H₄BrF₃O
Bod tání: 29,8…30…3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368691
CAS číslo: 886762-81-8
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR